Skip to Content
Merck
All Photos(1)

Key Documents

SMB00416

Sigma-Aldrich

Gentiopicroside

≥98% (HPLC)

Synonym(s):

(3S,4R)-4-ethenyl-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4,6-dihydro-3H-pyrano[3,4-c]pyran-8-one, (5R,6S)-5-Ethenyl-6-(-D-glucopyranosyloxy)-5,6-dihydro-1H,3H-Pyrano[3,4-c]pyran-1-one, (5R-trans)-5-ethenyl-6-(-D-glucopyranosyloxy)-5,6-dihydro-1H,3H-pyrano[3,4-c]pyran-1-one, Gentiopicrin, NSC606402

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$389.00

$389.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$389.00

About This Item

Empirical Formula (Hill Notation):
C16H20O9
CAS Number:
Molecular Weight:
356.32
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

$389.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

plant (Gentiana macrophylla Pall)

Quality Level

Assay

≥98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

C=C[C@H]1[C@H](O[C@]2([H])O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)OC=C3C(OCC=C31)=O

InChI

1S/C16H20O9/c1-2-7-8-3-4-22-14(21)9(8)6-23-15(7)25-16-13(20)12(19)11(18)10(5-17)24-16/h2-3,6-7,10-13,15-20H,1,4-5H2/t7-,10-,11-,12+,13-,15+,16+/m1/s1

InChI key

DUAGQYUORDTXOR-GPQRQXLASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Gentiopicroside (GPS) is a secoiridoid glycoside sourced from the dried roots of Gentiana rigescens.

Application

Gentiopicroside has been used as a reference standard for the quantification of iridoid glycoside in six Caucasian Gentians herb samples using high-performance liquid chromatography with diode array detection (HPLC-DAD) technique.

Biochem/physiol Actions

Gentiopicroside is a potent anti-arthritic agent. It displays hepatoprotective effects in arthritic rats by improving oxidative stress and reducing inflammation in combination with leflunomide and/or methotrexate by regulating mRNA genes and pro-inflammatory cytokines. It also displays its efficacy by inhibiting reactive oxygen species (ROS)-Nuclear factor-kappa B (NF-κB)-NOD-like receptor protein 3 (NLRP3) inflammasome axis in adjuvant-induced arthritis (AIA) in vitro and in vivo models. Gentiopicroside exerts antiaging properties in yeast prolonging lifespan and inducing mitophagy and reducing oxidative stress. It prevents osteoporosis by regulating β-catenin-BMPsignaling and promoting osteogenesis in vitro.
Gentiopicroside is an important active component of secoiridoid glycosides from Gentiana macrophylla Pall that exhibit anti-inflammatory, antibacterial and liver protecting activities. Gentiopicroside protects chondrocytes from IL-1β-induced inflammation response.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Zhijie Wan et al.
Life sciences, 265, 118689-118689 (2020-11-02)
This study aimed to examine whether gentiopicroside (GPS) could exert hepatoprotective effects on leflunomide (LEF)- and/or methotrexate (MTX)-treated arthritic rats through anti-inflammatory and antioxidant pathways. We observed the external symptoms of joints, analysed serum indicators, measured haematological parameters and mRNA
Huaji Jiang et al.
Journal of cellular and molecular medicine, 25(23), 10825-10836 (2021-11-17)
Osteoporosis is characterized by increased bone fragility, and the drugs used at present to treat osteoporosis can cause adverse reactions. Gentiopicroside (GEN), a class of natural compounds with numerous biological activities such as anti-resorptive properties and protective effects against bone
Na Jia et al.
Journal of ethnopharmacology, 189, 1-9 (2016-05-18)
The flowers of Gentiana macrophylla have been usually applied to cure the joint inflammation and rheumatoid arthritis in Traditional Chinese Medicine. This work aimed to investigate the anti-rheumatoid arthritic effect and possible mechanism of iridoid glycosides from G. macrophylla (GMI)
Daniil N Olennikov et al.
Metabolites, 9(11) (2019-11-11)
The members of Gentiana genus are widely distributed in the Caucasus region where they are used as phytoremedies, but they still have not been studied for their chemical composition and bioactivity. High-performance liquid chromatography with diode array and electrospray triple
Meiling Wang et al.
Frontiers in pharmacology, 11, 515-515 (2020-06-02)
Rheumatoid arthritis (RA) is among the most prevalent forms of autoimmunity. Gentiopicroside (Gent) is an iridoid glucoside derived from the Gentiana Macrophylla Pall which is used in traditional Chinese medicine to treat RA. The present study was designed to explore

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service