Skip to Content
Merck
All Photos(1)

Key Documents

SMB00195

Sigma-Aldrich

Maesopsin

≥90% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$560.00

$560.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
1 MG
$560.00

About This Item

Empirical Formula (Hill Notation):
C15H12O6
CAS Number:
Molecular Weight:
288.25
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

$560.00


Usually ships in 2 weeks.

Request a Bulk Order

Assay

≥90% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

Oc1ccc(CC2(O)Oc3cc(O)cc(O)c3C2=O)cc1

InChI

1S/C15H12O6/c16-9-3-1-8(2-4-9)7-15(20)14(19)13-11(18)5-10(17)6-12(13)21-15/h1-6,16-18,20H,7H2

InChI key

LOFYFDPXORJJEE-UHFFFAOYSA-N

General description

Natural product derived from plant source.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liselotte Krenn et al.
Journal of natural products, 66(8), 1107-1109 (2003-08-23)
A sulfated emodin glucoside, emodin 8-O-beta-D-glucopyranosyl-6-O-sulfate (1), was isolated from the roots of Rheum emodi in an investigation of the active constituents of this Nepalese medicinal plant, and its structure was determined by spectroscopic and chemical methods. Additionally, two rare
X C Li et al.
Phytochemistry, 46(1), 97-102 (1997-09-01)
During the search for antimicrobial compounds from higher plant sources, a methanol extract of Ceanothus americanus demonstrated antimicrobial activity against selected oral pathogens. Through further bioassay-guided fractionation and purification, three triterpenes (ceanothic acid, 27-hydroxy ceanothic acid and ceanothetric acid) and
R Bekker et al.
Journal of natural products, 64(3), 345-347 (2001-03-30)
Resolution of racemic 2-benzyl-2-hydroxy-1-benzofuran-3(2H)-ones (auronols) and CD data of the ensuing enantiomers permit assessment of the absolute configuration at the single stereocenter.
R Chao et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 22(4), 233-234 (1997-04-01)
A reversed-phase HPLC method was developed for the determination of maesopsin, a fliavonoid, in the xylem parts of the traditional Tibetan medicinal herb Shengdeng (Rhamnella gilgitica). An ODS column was used and the mobile phase was methanol-water (28:27) with 0.05

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service