Skip to Content
Merck
All Photos(1)

Key Documents

SMB00161

Sigma-Aldrich

Aceroside VIII

≥95% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$373.00

$373.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
1 MG
$373.00

About This Item

Empirical Formula (Hill Notation):
C30H42O12
CAS Number:
Molecular Weight:
594.65
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

$373.00


Usually ships in 2 weeks.

Request a Bulk Order

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

OC[C@]1(O)[C@@H](O)[C@@H](OC1)OC[C@H]([C@@H](O)[C@H](O)[C@H]2O)O[C@H]2O[C@H](CCCCC3=CC=C(O)C=C3)CCC4=CC=C(O)C=C4

InChI

1S/C30H42O12/c31-16-30(38)17-40-29(27(30)37)39-15-23-24(34)25(35)26(36)28(42-23)41-22(14-9-19-7-12-21(33)13-8-19)4-2-1-3-18-5-10-20(32)11-6-18/h5-8,10-13,22-29,31-38H,1-4,9,14-17H2/t22-,23-,24-,25+,26-,27+,28-,29-,30-/m1/s1

InChI key

JLMGCBFIPZDHLZ-GYOMMSMMSA-N

General description

Natural product derived from plant source.

Biochem/physiol Actions

Aceroside VIII acts as a specific histone deacetylase 6 (HDAC6) inhibitor.[1] It has an ability to synergistically enhance anticancer activity of other HDAC inhibitors in colon cancer cells.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aceroside VIII is a new natural selective HDAC6 inhibitor that synergistically enhances the anticancer activity of HDAC inhibitor in HT29 cells.
Ryu H W, et al.
Planta Medica, 81(03), 222-227 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service