Skip to Content
Merck
All Photos(1)

Documents

SMB00113

Sigma-Aldrich

6-Geranyl-7-hydroxycoumarin

≥95% (LC/MS-ELSD)

Synonym(s):

6-(3,7-Dimethyl-2,6-octadienyl)-7-hydroxycoumarin, Ostruthin, Ostruthine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22O3
CAS Number:
Molecular Weight:
298.38
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

C\C(C)=C\CC\C(C)=C\Cc1cc2C=CC(=O)Oc2cc1O

InChI

1S/C19H22O3/c1-13(2)5-4-6-14(3)7-8-15-11-16-9-10-19(21)22-18(16)12-17(15)20/h5,7,9-12,20H,4,6,8H2,1-3H3/b14-7+

InChI key

INBMTJJPUABOQJ-VGOFMYFVSA-N

General description

Natural product derived from plant source.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Qi et al.
Bioscience, biotechnology, and biochemistry, 63(8), 1501-1502 (1999-09-29)
A search for neuritogenic compounds in Chinese herbs resulted in the isolation of two known substances, ostruthin and (-)-bornyl ferulate, from Notopterygium incisum (and/or N. forbesii). Both compounds induced comparable neurite-like structures in 20% of rat PC12 cells at 2
Andreas Schinkovitz et al.
Planta medica, 69(4), 369-371 (2003-04-24)
Following a bioassay-guided fractionation, ostruthin (6-geranyl-7-hydroxycoumarin) was isolated from the roots of Peucedanum ostruthium Koch (Apiaceae) as a compound with pronounced in vitro activity against several species of rapidly growing Mycobacteria, namely Mycobacterium abscesus, M. aurum, M. fortuitum, M. phlei
Helge Joa et al.
Journal of natural products, 74(6), 1513-1516 (2011-06-02)
Inhibition of vascular smooth muscle cell (VSMC) proliferation is of substantial interest in combating cardiovascular disease. A dichloromethane extract from the rhizomes of Peucedanum ostruthium, a traditionally used Austrian medicinal plant with anti-inflammatory properties, was examined for a putative antiproliferative
Y Cai et al.
Carcinogenesis, 18(8), 1521-1527 (1997-08-01)
Several naturally occurring coumarins previously found to be potent inhibitors of mouse hepatic ethoxyresorufin-O-deethylase (EROD) and/or pentoxyresorufin-O-dealkylase (PROD) were examined for their effects on formation of benzo[a]pyrene (B[a]P) and 7,12-dimethylbenz[a]anthracene (DMBA) DNA adducts in mouse epidermis, as well as, their
Feng Li et al.
Planta medica, 78(8), 796-799 (2012-04-21)
During a search for potent anticancer agents from natural products based on an anti-austerity strategy, we found that a CHCl3 extract of Rhizoma et Radix Notopterygii (Qianghuo), a Chinese crude drug, exhibited strong cytotoxicity against PANC-1 human pancreatic cancer cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service