Skip to Content
Merck
All Photos(1)

Key Documents

SMB00073

Sigma-Aldrich

Bergenin

≥95% (LC/MS-ELSD)

Synonym(s):

Cuscutin, Ardisic acid B, Vakerin

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$427.00

$427.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
1 MG
$427.00

About This Item

Empirical Formula (Hill Notation):
C14H16O9
CAS Number:
Molecular Weight:
328.27
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

$427.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1[C@@H]2[C@H](OC(=O)c3c2c(c(c(c3)O)OC)O)[C@H]([C@@H]([C@H]1CO)O)O

InChI

1S/C14H16O9/c1-21-11-5(16)2-4-7(9(11)18)12-13(23-14(4)20)10(19)8(17)6(3-15)22-12/h2,6,8,10,12-13,15-19H,3H2,1H3/t6-,8-,10+,12+,13-/m1/s1

InChI key

YWJXCIXBAKGUKZ-HJJNZUOJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural product derived from plant source.

Biochem/physiol Actions

Bergenin displays antinociceptive and anti-inflammatory properties, in addition to its use in folk medicines as a potent hepatoprotective agent. Bergenin acts by inhibiting the formation of interleukin 1beta and TNF-α, which suggests its potential use for controling inflammatory pain.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong Wang et al.
Bioorganic & medicinal chemistry letters, 15(18), 4073-4075 (2005-07-05)
A novel C-C-coupled dimer derivative of bergenin was produced by the biotransformation of cultured mycelia of white rot fungus Pleurotus ostreatus. Its structure was elucidated by detailed spectroscopic analysis.
Heitor A De Abreu et al.
Organic & biomolecular chemistry, 6(15), 2713-2718 (2008-07-18)
(+)-Bergenin (1) was isolated from Sacoglottis uchi, a species of vegetable found in the Amazon region and popularly used for the treatment of several hepatic problems. The structure of 1 was fully characterized using IR, GC-MS and NMR (1D and
B Nyasse et al.
Die Pharmazie, 59(6), 492-494 (2004-07-14)
The presence of bergenin in substantial amounts in the methanol leaves extract of Flueggea virosa (Euphorbiaceae) was established as a strong chemotaxomic point of differentiation between Flueggea virosa and Securinega virosa. Bergenin showed an inhibitory effect on the growth of
Yan-Bin Shi et al.
Biomedical chromatography : BMC, 20(10), 1065-1070 (2006-04-04)
A validated reversed-phase high-performance liquid chromatographic (RP-HPLC) method was developed for the determination of bergenin in rat plasma. Bergenin in rat plasma was extracted with methanol, which also acted as a deproteinization agent. Chromatographic separation of bergenin was performed on
Shuang He et al.
Drug development and industrial pharmacy, 38(10), 1280-1288 (2011-12-31)
A novel coated gastric floating drug-delivery system (GFDDS) of bergenin (BN) and cetirizine dihydrochloride (CET) was developed. First, the pharmacodynamic studies were performed and the results revealed that the new compounds of bergenin/cetirizine dihydrochloride had comparative efficacy as commercial products

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service