Skip to Content
Merck
All Photos(1)

Key Documents

SMB00055

Sigma-Aldrich

Benzomalvin-C

≥95% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$866.00

$866.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
1 MG
$866.00

About This Item

Empirical Formula (Hill Notation):
C24H17N3O3
CAS Number:
Molecular Weight:
395.41
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

$866.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

CN1C(=O)c2ccccc2N3C(=O)c4ccccc4N=C3[C@]15O[C@H]5c6ccccc6

InChI

1S/C24H17N3O3/c1-26-21(28)17-12-6-8-14-19(17)27-22(29)16-11-5-7-13-18(16)25-23(27)24(26)20(30-24)15-9-3-2-4-10-15/h2-14,20H,1H3/t20-,24-/m0/s1

InChI key

TWDKBDSVUUKABK-RDPSFJRHSA-N

General description

Natural product derived from fungal source.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H H Sun et al.
The Journal of antibiotics, 47(5), 515-522 (1994-05-01)
In the course of screening microbial broths for neurokinin receptor antagonists, a series of new benzodiazepines, benzomalvins A (1), B (2) and C (3), has been isolated from the culture broth of a fungus identified as a Penicillium sp. Benzomalvin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service