Skip to Content
Merck
All Photos(3)

Documents

SILA200

Sigma-Aldrich

Silicic acid

suitable for column chromatography, 60-200 mesh

Synonym(s):

Metasilicic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H2O3Si
CAS Number:
Molecular Weight:
78.10
EC Number:
MDL number:
UNSPSC Code:
23201100
PubChem Substance ID:
NACRES:
NA.22

particle size

60-200 mesh

suitability

suitable for column chromatography

SMILES string

O[Si](O)=O

InChI

1S/H2O3Si/c1-4(2)3/h1-2H

InChI key

IJKVHSBPTUYDLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For resolution of acidic mixtures

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher Exley et al.
Journal of Alzheimer's disease : JAD, 10(1), 17-24 (2006-09-22)
There are unexplained links between human exposure to aluminium and the incidence, progression and aetiology of Alzheimer's disease. The null hypothesis which underlies any link is that there would be no Alzheimer's disease in the effective absence of a body
A handbook for uncovering the complete energetic budget in insects: the van Handel's method (1985) revisited.
Foray V,et al.
Physiological Entomology, 37(3), 295-302 (2012)
M J González-Muñoz et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 46(1), 49-56 (2007-08-19)
Aluminium (Al), a neurotoxin, has lately been implicated as one of the possible causal factors contributing to Alzheimer's disease. Because silicon (Si) intake can affect the bioavailability of aluminium, the object of the present study was to assess whether moderate
Hiroaki Saito et al.
Journal of oleo science, 64(6), 603-616 (2015-06-02)
The lipid and fatty acid composition of the muscle of the wild Japanese freshwater eel, Anguilla japonica, was analyzed between the initial and terminal stages of spawning migration to clarify the relationship between lipid physiology and maturation. Triacylglycerols were the
Hideshi Nakamura et al.
International journal of pharmaceutics, 493(1-2), 305-312 (2015-07-29)
In this study, the human gustatory palatability sensation of taste-masked famotidine and amlodipine orally disintegrating tablets (ODTs) was quantitatively predicted by an electronic gustatory system (α-Astree e-Tongue). Furthermore, its use in formulation design was evaluated. The famotidine- and amlodipine-containing ODTs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service