Skip to Content
Merck
All Photos(1)

Key Documents

SCP0244

Sigma-Aldrich

Substance P Peptide

≥95% (HPLC), lyophilized, Sar⁹

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$147.00
5 MG
$506.00

$147.00


Usually ships in 2 weeks.


Select a Size

Change View
1 MG
$147.00
5 MG
$506.00

About This Item

Empirical Formula (Hill Notation):
C64H100N18O13S1
Molecular Weight:
1361.66
UNSPSC Code:
12352209
NACRES:
NA.32

$147.00


Usually ships in 2 weeks.

Product Name

[Sar9]-Substance P,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥72%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Arg-Pro-Lys-Pro-Gln-Gln-Phe-Phe-Sar-Leu-Met-NH2

Application

Substance P is a tachykinin family 11 amino acid neuropeptide involved in neurotransmission and neuromodulation. Substance P acts via the neurokinin-1 receptor (NK1-receptor, NK1R). [Sar9]-Substance P and [Sar9] substance P-sulphone (NK1-receptor agonist) are used to study and differentiate NK1-receptors activities from NK2R and NK3R activities.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service