Skip to Content
Merck
All Photos(1)

Key Documents

SCP0217

Sigma-Aldrich

PPlases Substrate, fluorogenic

≥95% (HPLC), lyophilized

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$164.00

$164.00


Usually ships in 2 weeks.


Select a Size

Change View
10 MG
$164.00

About This Item

Empirical Formula (Hill Notation):
C37H44N5O9
Molecular Weight:
702.77
UNSPSC Code:
12352204
NACRES:
NA.32

$164.00


Usually ships in 2 weeks.

Product Name

PPlases Substrate, fluorogenic,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥80%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Suc-Ala-Leu-Pro-Phe-AMC

Application

Suc-Ala-Leu-Pro-Phe-pNA, colorimetric and Suc-Ala-Leu-Pro-Phe-AMC, fluorogenic are used as substrates for peptidyl-prolyl cis/trans isomerases (PPIases, EC: 5.2.1.8).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. What is the maximum excitation and emssion wavelength for Product SCP0217, PPlases Substrate, fluorogenic?

    1 answer
    1. The maximum excitation wavelength for Product SCP0217, PPlases Substrate, fluorogenic is 360-380 nm and the emission wavelength is 440-460nm.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service