Skip to Content
Merck
All Photos(1)

Key Documents

SCP0200

Sigma-Aldrich

Neuromedin N

Synonym(s):

NMN-125, NN, NT/N, NmN

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$253.00

$253.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
10 MG
$253.00

About This Item

Empirical Formula (Hill Notation):
C38H63N7O8
Molecular Weight:
745.95
UNSPSC Code:
12352200
NACRES:
NA.32

$253.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥65%

storage condition

protect from light

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... NTS(4922)

Amino Acid Sequence

Lys-Ile-Pro-Tyr-Ile-Leu

Biochem/physiol Actions

Neuromedin N is a hexapeptide that is related to neurotensin.[1] Its precursor, proneuromedin N, is expressed in the adrenals, brain, gut and also in several cancers.[2] In myoelectric activity, it has been reported to act in a similar manner as neurotensin, at a lesser degree of potency. Neuromedin N has been shown to increase the chemotaxis capacity of mouse lymphocytes and to modulate function of the hypothalamo-pituitary-adrenocortical axis in rats. Neuromedin N has been proposed to exert its actions via increase in intracellular Ca2+ concentration, although this is uncertain.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Investigation of proNT/NMN secretion from small cell lung carcinoma cells using a mouse xenograft model.
Wakabayashi-Nakao K
Oncology Reports, 28(4), 1181-1186 (2012)
L K Malendowicz et al.
Experimental and toxicologic pathology : official journal of the Gesellschaft fur Toxikologische Pathologie, 47(1), 41-45 (1995-01-01)
The effects of a subcutaneous bolus injection of 2 micrograms neurotensin (NT) or neuromedin N (NMN) on the function of the hypothalamo-pituitary-adrenocortical axis were investigated in both normal and cold-stressed rats. The blood concentrations of ACTH, corticosterone (B) and aldosterone
J J Garrido et al.
Regulatory peptides, 41(1), 27-37 (1992-09-03)
The action of neurotensin and related peptides has not been yet studied on lymphocytes, although there are studies indicating the stimulative action of neurotensin, a peptide first isolated from bovine hypothalamus, on different functions of phagocytic immune cells. The present
Neuromedin N: high affinity interaction with brain neurotensin receptors and rapid inactivation by brain synaptic peptidases.
Checler F
European Journal of Pharmacology, 126(3), 239-244 (1986)
I Schultz et al.
Regulatory peptides, 35(3), 197-205 (1991-09-03)
The effects of neuromedin-N on migrating myoelectric complexes in the small intestine of rats were studied. As neuromedin-N and neurotensin are structurally related peptides a comparison with neurotensin was made. Myoelectric activity was recorded by means of three bipolar electrodes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service