Skip to Content
Merck
All Photos(1)

Key Documents

SCP0171

Sigma-Aldrich

Interleukin 1b Converting Enzyme Substrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H36N6O10
Molecular Weight:
628.63
UNSPSC Code:
12352200
NACRES:
NA.32

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥75%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Ac-Tyr-Val-Ala-Asp-pNA

Application

Ac-YVAD-pNa (acetyl-Tyr-Val-Ala-Asp-p-nitroanalide) is a selective chromogenic substrate for the detection of Interleukin-1b Converting Enzyme/caspase 1.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Bossenmeyer-Pourié et al.
Neuroscience, 95(4), 1157-1165 (2000-02-22)
To further explore the contribution of caspase-1/interleukin-1beta-convening enzyme in the consequences of hypoxia in developing brain neurons, its temporal expression profile was analysed by immunohistochemistry and western blotting in cultured neurons from the embryonic rat forebrain subjected to a hypoxic
Ori Nov et al.
Endocrinology, 151(9), 4247-4256 (2010-07-28)
Central obesity is frequently associated with adipose tissue inflammation and hepatic insulin resistance. To identify potential individual mediators in this process, we used in vitro systems and assessed if insulin resistance in liver cells could be induced by secreted products
Natasha Ann Pereira et al.
Biochemical and biophysical research communications, 377(3), 873-877 (2008-11-04)
Many designated substrates and inhibitors have been widely used to investigate the roles of caspases in apoptotic death during mammalian cell culture. However, the specificities of these substrates and inhibitors have not been systematically evaluated. As a result, conclusions on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service