Skip to Content
Merck
All Photos(1)

Key Documents

SCP0142

Sigma-Aldrich

ET Antag PD 156252

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
$182.00
1 MG
$258.00

$182.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
500 μG
$182.00
1 MG
$258.00

About This Item

Empirical Formula (Hill Notation):
C53H69N7O10
Molecular Weight:
964.16
UNSPSC Code:
12352200
NACRES:
NA.32

$182.00


Usually ships in 2 weeks.

Request a Bulk Order

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥85%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Ac-Bhg-Leu-Asp-Ile-NMeIle-Trp

Application

PD-156252 is used as an endothelin-A/endothelin-B (ETA/ETB) receptor antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Javed Iqbal et al.
Mini reviews in medicinal chemistry, 5(4), 381-408 (2005-04-28)
Endothelins (ETs) are potent vasoconstrictor peptides and are associated with several disease states like pulmonary hypertension, systemic hypertension and heart failure. Endothelin-1 (ET-1) is the first member of the family and it has the receptor subtypes known as ETA and
Pierre Shephard et al.
Thrombosis and haemostasis, 92(2), 262-274 (2004-07-23)
Myofibroblasts are specialized fibroblasts that contribute to wound healing by producing extracellular matrix and by contracting the granulation tissue. They appear in a phase of wound healing when the dermis strongly interacts with activated epidermal keratinocytes. Direct co-culture with keratinocytes
W L Cody et al.
Journal of medicinal chemistry, 40(14), 2228-2240 (1997-07-04)
The endothelins (ETs) are a family of bicyclic 21-amino acid peptides that are potent and prolonged vasoconstrictors. It has been shown that highly potent combined ETA/ETB receptor antagonists can be developed from the C-terminal hexapeptide of ET (His16-Leu17-Asp18-Ile19-Ile20-Trp21), such as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service