Skip to Content
Merck
All Photos(1)

Key Documents

SCP0075

Sigma-Aldrich

Caspase 13 Substrate

≥95% (HPLC), lyophilized

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$147.00

$147.00


Usually ships in 2 weeks.


Select a Size

Change View
5 MG
$147.00

About This Item

Empirical Formula (Hill Notation):
C28H38N6O13
Molecular Weight:
666.63
UNSPSC Code:
12352202
NACRES:
NA.32

$147.00


Usually ships in 2 weeks.

Product Name

Caspase 13 Substrate,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥80%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Ac-Leu-Glu-Glu-Asp-pNA

Application

Caspase 13 (ERICE), an orthologue of human caspase 4, is a cysteine protease which may be detected using a variety of chromogenic and fluorogenic peptide substrates built around the LEED (Leu-Glu-Glu-Asp) sequence. These substrates include: Ac-LEED-pNa (acetyl-Leu-Glu-Glu-Asp-p-nitroanalide), chromogenic and Ac-LEED-AMC (Acetyl-Leu-Glu-Glu-Asp- amino-4-methylcoumarin), fluorogenic.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U Koenig et al.
Biochemical and biophysical research communications, 285(5), 1150-1154 (2001-08-02)
Caspase-13 was reported to be a member of the human caspase family of proteases (Humke, E., et al., J. Biol. Chem. 273, 15702-15707, 1998). By contrast, a recent study (Lin, X., et al., J. Biol. Chem. 275, 39920-39926, 2000) could
S Taylor et al.
DNA sequence : the journal of DNA sequencing and mapping, 10(6), 387-394 (2000-05-29)
Caspases are cysteine proteases which have important roles in the activation of cytokines and in apoptosis. The ICE subfamily of caspases comprise peptides closely related to caspase-1, or interleukin-1beta (IL-1beta) converting enzyme (ICE), which promotes maturation of interleukin IL-1beta and
E W Humke et al.
The Journal of biological chemistry, 273(25), 15702-15707 (1998-06-23)
Programmed cell death, or apoptosis, is a process of fundamental importance to cellular homeostasis in metazoan organisms (Ellis, R. E., Yuan, J., and Horvitz, H. R. (1991) Annu. Rev. Cell Biol. 7, 663-698). The caspase family of mammalian proteases, related

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service