Skip to Content
Merck
All Photos(1)

Key Documents

SCP0045

Sigma-Aldrich

Amyloid β 27-35 human

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$110.00
5 MG
$381.00

$110.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
1 MG
$110.00
5 MG
$381.00

About This Item

Empirical Formula (Hill Notation):
C40H73N11O11S1
Molecular Weight:
916.14
UNSPSC Code:
12352200
NACRES:
NA.32

$110.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥68%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met

Application

Amyloid β (Aβ) refers to peptides derived from Amyloid precursor protein that vary in length from 36-43 amino acids. Aβ(s) peptides, their peptide fragments and mutated fragments are used to study a wide range of metabolic and regulatory functions including activation of kinases, regulation of cholesterol transport, function as a transcription factor, and regulators of inflammation. Aβ(s) peptides and their peptide fragments are also used to study oxidative stress, metal binding and mechanisms of protein cross-linking in the context of diseases such as Alzheimer′s disease and neurodegeneration.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wah Chin Boon et al.
Behavioural brain research, 209(1), 27-35 (2010-01-21)
The aim of this study was to characterize APPC100.V717F transgenic (TgC100.V717F) mice which over-express a mutant C100 fragment of the amyloid precursor protein. The mice were compared to TgC100 wild type mice (TgC100.WT) and non-transgenic controls at 4-9 and 16-22

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service