Skip to Content
Merck
All Photos(1)

Documents

SCP0007

Sigma-Aldrich

ADAM substrate

≥95% (HPLC), lyophilized

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C64H85N13O18S2
Molecular Weight:
1388.57
UNSPSC Code:
12352202
NACRES:
NA.32

product name

ADAM substrate, ≥95% (HPLC)

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥70%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

NMe-Phe-Lys-Pro-Ala-Lys-Phe-Phe-Arg-Leu-Dabcyl-Lys-NH2

Application

ADAM substrate containing the fluorophore quencher Dabcy is used in FRET applications for detection of metalloprotease activity. DABCYL is a popular acceptor for developing FRET-based protease substrates. DABCYL dyes are often paired with EDANS in FRET-based fluorescent probes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcia L Moss et al.
Analytical biochemistry, 366(2), 144-148 (2007-06-06)
In this paper we describe novel fluorescent substrates for the human ADAM family members ADAM17, ADAM10, ADAM8, and ADAM12 that have good specificity constants and are useful for high-throughput screening of inhibitors. The fluorescence resonance energy transfer substrates contain a
Marcia L Moss et al.
Combinatorial chemistry & high throughput screening, 13(4), 358-365 (2009-12-18)
Fluorescence resonance energy transfer substrates were designed and tested as substrates for ADAM9. The donor/quencher pair used were 5-carboxy fluorescein (Fam) and 4-(4-dimethyl-aminophenylazo)benzoyl (Dabcyl) since they have been well studied sensitive fluorescent probes. The peptides based on precursor TNF-alpha, Dabcyl-Ser-Pro-Leu-Ala-Gln-Ala-Val-Arg-Ser-Ser-Lys(Fam)-
Characterization of Mca-Lys-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH2, a fluorogenic substrate with increased specificity constants for collagenases and tumor necrosis factor converting enzyme.
Neumann U, Kubota H, Frei K, et al.
Analytical Biochemistry, 328, 166-173 (328)
Bernhard F Becker et al.
Biological chemistry, 383(11), 1821-1826 (2003-01-18)
Tumor necrosis factor-alpha (TNFalpha) is presumably shed from cell membranes by TNFalpha-cleaving enzyme (TACE). The peptides SPLAQAVRSSSR and Dabcyl-LAQAVRSSSR-Edans, each encompassing the cleavage sequence of pro-TNFalpha recognized by TACE, were applied to intact umbilical vein endothelium (HUVEC), peripheral blood leukocytes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service