Skip to Content
Merck
All Photos(2)

Key Documents

SBR00050

Sigma-Aldrich

FITC Labeled D-Lysine

Suitable for bacterial labeling

Synonym(s):

FADL, FDAA

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$312.00

$312.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$312.00

About This Item

Empirical Formula (Hill Notation):
C27H25N3O7S
Molecular Weight:
535.57
UNSPSC Code:
12352209
NACRES:
NA.24

$312.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

storage temp.

−20°C

General description

Bacterial Peptidoglycan is a polymer consisting of sugars and amino acids, a mesh-like rigid layer that forms the cell wall. D-Amino Acids (D-AAs) are metabolically incorporated onto the bacterial cell wall by D,D-transpeptidase and/or L,D-transpeptidase. It was recently discovered that those transpeptidases catalyze the metabolic incorporation of exogenous D-AAs with almost no restriction of the side-chain identity.[1] Modification of a D-AA with molecular fluorophores provides fluorescent D-amino acids that can efficiently label in situ peptidoglycan in diverse bacterial species.[2]
Fluorescein Labeled D-Lysine is a fluorescent derivative of D-Lysine that is obtained by a synthetic conjugation of D-Lysine to a fluorophore. Fluorescein Labeled D-Lysine has high biocompatibility and suitability for labeling peptidoglycans in live bacteria. Additionally, it can be used in tandem with other stains such as: FITC-labelled Alanine, Rhodamine B labelled Polymyxin B, FITC labelled vancomycin and Dansyl labelled polymyxin B (SBR00049, SBR00036, SBR00028, SBR00029).

Application

Fluorescent labeled D-AAs can be used for many applications including:
  • Bacterial cell wall morphology
  • Bacterial cell wall formation or remodeling activity
  • Bacterial viability/activity
  • Identify bacterial activity on surfaces or in substances
  • Differentiation betweenvarious bacterial strains according to their incorporation profile of differentD amino acids and sugars
[3]

Analysis Note

  • Fluorescent microscopy application: Fluorescein Labeled D-Lysine has excitation/emission wavelength range at 495 nm/515 nm.
  • The recommended working concentration in fluorescent microscopy imaging application is between 250μM-500 μM in DMSO.
  • Aliquots of the DMSO solution can be stored at -20°C, protected from light for at least one month.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helene Botella et al.
mBio, 8(5) (2017-09-14)
Peptidoglycan (PG), a polymer cross-linked by d-amino acid-containing peptides, is an essential component of the bacterial cell wall. We found that a fluorescent d-alanine analog (FDAA) incorporates chiefly at one of the two poles in Mycobacterium smegmatis but that polar
Yen-Pang Hsu et al.
Chemical science, 8(9), 6313-6321 (2017-10-11)
Fluorescent d-amino acids (FDAAs) enable efficient in situ labeling of peptidoglycan in diverse bacterial species. Conducted by enzymes involved in peptidoglycan biosynthesis, FDAA labeling allows specific probing of cell wall formation/remodeling activity, bacterial growth and cell morphology. Their broad application
Felipe Cava et al.
The EMBO journal, 30(16), 3442-3453 (2011-07-28)
Production of non-canonical D-amino acids (NCDAAs) in stationary phase promotes remodelling of peptidoglycan (PG), the polymer that comprises the bacterial cell wall. Impairment of NCDAAs production leads to excessive accumulation of PG and hypersensitivity to osmotic shock; however, the mechanistic
Erkin Kuru et al.
Angewandte Chemie (International ed. in English), 51(50), 12519-12523 (2012-10-12)
Tracking a bug's life: Peptidoglycan (PG) of diverse bacteria is labeled by exploiting the tolerance of cells for incorporating different non-natural D-amino acids. These nontoxic D-amino acids preferably label the sites of active PG synthesis, thereby enabling fine spatiotemporal tracking

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service