Skip to Content
Merck
All Photos(1)

Key Documents

SBR00011

Sigma-Aldrich

Clarithromycin, Ready Made Solution

50 mg/mL in DMSO

Synonym(s):

6-O-methyl erythromycin

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
$329.00

$329.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
2 ML
$329.00

About This Item

Empirical Formula (Hill Notation):
C38H69NO13
CAS Number:
Molecular Weight:
747.95
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85

$329.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

Ready Made Solution

Quality Level

form

liquid

storage condition

dry media (Keep container tightly closed in a dry and well-ventilated place. Storage class (TRGS 510) Combustible liquids.)

concentration

50 mg/mL in DMSO

color

colorless to faint yellow

antibiotic activity spectrum

Gram-positive bacteria

Mode of action

protein synthesis | interferes

storage temp.

−20°C

SMILES string

N([C@@H]1[C@H]([C@@H](O[C@@H](C1)C)O[C@@H]2[C@H]([C@@H]([C@H](C(=O)O[C@@H]([C@]([C@@H]([C@H](C(=O)[C@@H](C[C@]2(OC)C)C)C)O)(O)C)CC)C)O[C@@H]3O[C@H]([C@@H]([C@@](C3)(OC)C)O)C)C)O)(C)C

InChI

1S/C38H69NO13/c1-15-26-38(10,45)31(42)21(4)28(40)19(2)17-37(9,47-14)33(52-35-29(41)25(39(11)12)16-20(3)48-35)22(5)30(23(6)34(44)50-26)51-27-18-36(8,46-13)32(43)24(7)49-27/h19-27,29-33,35,41-43,45H,15-18H2,1-14H3/t19-,20-,21+,22+,23-,24+,25+,26-,27+,29-,30+,31-,32+,33-,35+,36-,37-,38-/m1/s1

InChI key

AGOYDEPGAOXOCK-KCBOHYOISA-N

General description

Clarithromycin is a macrolide antibiotic, which has both antibacterial and anti-inflammatory actions. It is a semi-synthetic 14-membered derivative of erythromycin A, which is resistant to degradation under acidic conditions.[1][2]

Application

Clarithromycin may be used for-
  • the study of the effects of clarithromycin on rate-dependent changes in QT, Tp-e and proarrhythmic events performed on arterially perfused rabbit left ventricular wedge preparations[3]
  • the culture of parasites expressing GFP (green fluorescence protein) targeted to the apicoplast, to be further used for microscopy-based assays[4]
  • the study of the synergism of NAB741 with clarithromycin in microtiter plates.[5]

Biochem/physiol Actions

Macrolides, such as clarithromycin, prevent protein synthesis in bacteria, by binding to 50S ribosomal subunit. It also binds to other ribosomal proteins, and prevents the translocation of pepti-dyl-tRNA.[2] In patients with refractory asthma, it is capable of regulating the levels of interleukin-8 (IL-8), and neutrophil accumulation and activation in lungs. Thus, it might be used as an additional therapy in asthma to reduce noneosinophilic airway inflammation.[1]

Packaging

2ML

Storage and Stability

Keep container tightly closed in a dry and well-ventilated place. Storage class (TRGS 510) Combustible liquids.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Storage class (TRGS 510): Combustible liquids

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jodie L Simpson et al.
American journal of respiratory and critical care medicine, 177(2), 148-155 (2007-10-20)
Patients with refractory asthma have persistent symptoms despite maximal treatment with inhaled corticosteroids and long-acting bronchodilators. The availability of add-on therapies is limited, and effective add-on therapies that target noneosinophilic airway inflammation are needed. Macrolide antibiotics, such as clarithromycin, have
Shaobin Jia et al.
British journal of pharmacology, 164(2), 308-316 (2010-12-25)
Drug-induced torsades de pointes (TdP) often occurs during bradycardia due to reverse use-dependence. We tested the hypothesis that inhibition or enhancement of late sodium current (I(Na,L) ) could modulate the drug-induced reverse use-dependence in QT and T(p-e) (an index of
Eric H Ekland et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 25(10), 3583-3593 (2011-07-13)
Malarial parasites have evolved resistance to all previously used therapies, and recent evidence suggests emerging resistance to the first-line artemisinins. To identify antimalarials with novel mechanisms of action, we have developed a high-throughput screen targeting the apicoplast organelle of Plasmodium
Martti Vaara et al.
Antimicrobial agents and chemotherapy, 54(8), 3341-3346 (2010-05-19)
Polymyxins are cationic lipopeptides (five cationic charges) and the last resort for the treatment of serious Gram-negative infections caused by multiresistant strains. NAB741 has a cyclic peptide portion identical to that of polymyxin B but carries in the linear peptide
T Mazzei et al.
The Journal of antimicrobial chemotherapy, 31 Suppl C, 1-9 (1993-03-01)
After the discovery of erythromycin and other natural compounds, including oleandomycin, spiramycin, josamycin and midecamycin, much research has been devoted to synthesizing derivatives or analogues with improved chemical, biological and pharmacokinetic properties. These new macrolides are semisynthetic molecules that differ

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service