Skip to Content
Merck
All Photos(1)

Key Documents

SAE0195

Sigma-Aldrich

IceTEV Protease

Histidine tagged recombinant protein, aqueous solution

Synonym(s):

TEV protease

Sign Into View Organizational & Contract Pricing

Select a Size

1 KU
$220.00
10 KU
$1,100.00

$220.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 KU
$220.00
10 KU
$1,100.00

About This Item

UNSPSC Code:
12352204
NACRES:
NA.54

$220.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Quality Level

form

aqueous solution

shipped in

wet ice

storage temp.

−20°C

General description

TEV protease is a highly sequence specific serine protease from Tobacco Etch Virus. Its high specificity makes TEV protease popular for cleaving recombinant fusion proteins. However, at low temperatures such as 4 °C, standard TEV Protease has only ~10% of its activity compared to regular temperature conditions (such as 25-30 °C). IceTEV Protease is a new TEV Protease version that is designed to cleave fusion proteins effectively at low temperatures also, in the range of 2-10 °C, with comparable efficiency to cleavage by standard TEV Protease at 25-30 °C. Following the cleavage reaction, IceTEV Protease can be removed from the reaction mixture with nickel-agarose (Ni-agarose) beads.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Proteases for biotinylated tag removal for protein purification workflows with related reagents and technical resources.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service