Skip to Content
Merck
All Photos(1)

Key Documents

SAE0058

Sigma-Aldrich

Oxyrase® for Agar

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$122.00
100 ML
$216.00

$122.00


Estimated to ship onMay 18, 2025



Select a Size

Change View
50 ML
$122.00
100 ML
$216.00

About This Item

UNSPSC Code:
41106212
NACRES:
NA.54

$122.00


Estimated to ship onMay 18, 2025


storage temp.

−20°C

Quality Level

General description

Oxyrase® for Agar (OA is a sterile enzyme formulation that utilizes the Oxyrase®Enzyme System to produce anaerobic conditions in your custom agar. Oxyrase® For Agar and the Oxydish make isolating and growing anaerobes straight forward.

Legal Information

OxyDish is a trademark of Oxyrase, Inc.
Oxyrase is a registered trademark of Oxyrase, Inc.

accessory

Product No.
Description
Pricing

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service