Skip to Content
Merck
All Photos(1)

Key Documents

SAB5703008

Sigma-Aldrich

Anti-FOXF1 Antibody, clone 8F6G3, Rabbit Monoclonal

Synonym(s):

ACDMPV, FKHL5, FREAC1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
clone:
8F6G3, monoclonal
application:
WB
species reactivity:
mouse, mouse
technique(s):
western blot: 1:500 - 1:2000
citations:
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

material

colorless

clone

8F6G3, monoclonal

form

liquid

mol wt

40 kDa

species reactivity

mouse, mouse

concentration

1.07 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

MSSAPEKQQPPHGGGGGGGGGGGAAMDPASSGPSKAKKTNAGIRRPEKPPYSYIALIVMAIQSSPTKRLTLSEIYQFLQSRFPFFRGSYQGWKNSVRHNL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FOXF1(2294)

General description

This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the regulation of pulmonary genes as well as embryonic development. [provided by RefSeq, Jul 2008]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1-100 of human FOXF1 (Q12946).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service