Skip to Content
Merck
All Photos(4)

Key Documents

SAB5702212

Sigma-Aldrich

Anti-UCP1+UCP3 Antibody, clone 0S10O10, Rabbit Monoclonal

Synonym(s):

AI385626, Slc25a7, Ucp

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

0S10O10, monoclonal

form

liquid

mol wt

30 kDa

species reactivity

rat, mouse

concentration

1.00 mg/mL

technique(s)

immunofluorescence: 1:50 - 1:200
immunohistochemistry: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

MVNPTTSEVQPTMGVKIFSAGVSACLADIITFPLDTAKVRLQIQGEGQASSTIRYKGVLGTITTLAKTEGLPKLYSGLPAGIQRQISFASLRIGLYDSVQEYFSSGRETPASLGNKISAGLMTGGVAVFIGQPTEVVKVRMQAQSHLHGIKPRYTGTYNAYRVIATTESLSTLWKGTTPNLMRNVIINCTELVTYDLMKGALVNNKILADDVPCHLLSALVAGFCTTLLASPVDVVKTRFINSLPGQYPSVPSCAMSMYTKEGPTAFFKGFVASFLRLGSWNVIMFVCFEQLKKELMKSRQTVDCTT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... UCP1(22227)

General description

Mitochondrial protein responsible for thermogenic respiration, a specialized capacity of brown adipose tissue and beige fat that participates in non-shivering adaptive thermogenesis to temperature and diet variations and more generally to the regulation of energy balance. Functions as a long-chain fatty acid/LCFA and proton symporter, simultaneously transporting one LCFA and one proton through the inner mitochondrial membrane. However, LCFAs remaining associated with the transporter via their hydrophobic tails, it results in an apparent transport of protons activated by LCFAs. Thereby, dissipates the mitochondrial proton gradient and converts the energy of substrate oxydation into heat instead of ATP. Regulates the production of reactive oxygen species/ROS by mitochondria.

Immunogen

PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.
Recombinant fusion protein containing a sequence corresponding to amino acids 1-307 of mouse UCP1+UCP3 (P12242).

Application

WB, IHC, IF/ICC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service