SAB5702045
Anti-CR2/CD21 Antibody, clone 9F8A1, Rabbit Monoclonal
Synonym(s):
C3DR, CD21, CR, CVID7, SLEB9
About This Item
biological source
rabbit
Quality Level
material
colorless
clone
9F8A1, monoclonal
form
liquid
mol wt
155 kDa
species reactivity
rat, human
concentration
1.19 mg/mL
technique(s)
immunofluorescence: 1:50 - 1:200
immunohistochemistry: 1:50 - 1:200
western blot: 1:500 - 1:2000
color
colorless
isotype
IgG
immunogen sequence
GAVVTLECEDGYMLEGSPQSQCQSDHQWNPPLAVCRSRSLAPVLCGIAAGLILLTFLIVITLYVISKHRARNYYTDTSQKEAFHLEAREVYSVDPYNPAS
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... CR2(1380)
General description
Immunogen
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Skin Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service