Skip to Content
Merck
All Photos(5)

Key Documents

SAB5701141

Sigma-Aldrich

Anti-Pan Lactic acid-Lysine antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$346.00

$346.00


Estimated to ship onApril 04, 2025



Select a Size

Change View
100 μL
$346.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$346.00


Estimated to ship onApril 04, 2025


biological source

rabbit

Quality Level

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

mouse, rat, human

concentration

0.545 mg/mL

technique(s)

western blot: 1:500-1:2000

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

lactylation (Pan Lys)

Immunogen

A synthetic peptide of human Pan Lactic acid-Lysine.

Physical form

Buffer: PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service