Skip to Content
Merck
All Photos(4)

Key Documents

SAB5701126

Sigma-Aldrich

Anti-CDKN2A/p16INK4a antibody produced in mouse

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$346.00

$346.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZMS1072


Select a Size

Change View
100 μL
$346.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$346.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZMS1072

biological source

mouse

Quality Level

antibody product type

primary antibodies

clone

monoclonal

form

liquid

species reactivity

all

concentration

1.51 mg/mL

technique(s)

western blot: 1:500-1:2000

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CDKN2A(1029)

General description

This gene generates several transcript variants which differ in their first exons. At least three alternatively spliced variants encoding distinct proteins have been reported, two of which encode structurally related isoforms known to function as inhibitors of CDK4 kinase. The remaining transcript includes an alternate first exon located 20 Kb upstream of the remainder of the gene; this transcript contains an alternate open reading frame (ARF) that specifies a protein which is structurally unrelated to the products of the other variants. This ARF product functions as a stabilizer of the tumor suppressor protein p53 as it can interact with, and sequester, the E3 ubiquitin-protein ligase MDM2, a protein responsible for the degradation of p53. In spite of the structural and functional differences, the CDK inhibitor isoforms and the ARF product encoded by this gene, through the regulatory roles of CDK4 and p53 in cell cycle G1 progression, share a common functionality in cell cycle G1 control. This gene is frequently mutated or deleted in a wide variety of tumors, and is known to be an important tumor suppressor gene.

Immunogen

Recombinant protein of human CDKN2A/p16INK4a.

Physical form

Tris Buffer, pH7.3-7.7, with 1% BSA and <0.1% Sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service