SAB5600096
Anti-His-Tag Chimeric antibody, Human Monoclonal
recombinant, expressed in HEK 293 cells, clone RMH01, purified immunoglobulin
Synonym(s):
Anti-His tag antibody
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
Recommended Products
recombinant
expressed in HEK 293 cells
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
RMH01, monoclonal
recombinant monoclonal
form
buffered aqueous glycerol solution
technique(s)
capture ELISA: 0.1-2 μg/mL
western blot: 0.1-1 μg/mL
isotype
IgG
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
General description
Hexahistidine tag (His-tag) consists of four to six histidines fused genetically. It is widely used to purify proteins by immobilized metal-ion affinity chromatography (IMAC).
Specificity
This antibody is a Human-Rabbit chimeric antibody, made by fusion of the antigen binding region (variable domains of the heavy and light chains, VH and VL) of rabbit anti-HisTag monoclonal antibody (RM146), with the constant domain of Human IgG1. The antibody is recognized only by an anti-Human IgG secondary antibody.
Immunogen
Mixture of a peptide with 6xHis-Tag at the N-terminus and another peptide with 6xHis-Tag at the C-terminus.
Features and Benefits
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Physical form
Solution in phosphate buffered saline containing 50% glycerol, 1% BSA and 0.09% sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jonathan F Lovell et al.
BMC medicine, 20(1), 462-462 (2022-11-30)
Numerous vaccine strategies are being advanced to control SARS-CoV-2, the cause of the COVID-19 pandemic. EuCorVac-19 (ECV19) is a recombinant protein nanoparticle vaccine that displays the SARS-CoV-2 receptor-binding domain (RBD) on immunogenic nanoliposomes. Initial study of a phase 2 randomized
Tandem immobilized metal-ion affinity chromatography/immunoaffinity purification of His-tagged proteins?evaluation of two anti-His-tag monoclonal antibodies
Muller KM, et al.
Analytical Biochemistry, 259(1), 54-61 (1998)
Loree Heller et al.
Journal of cellular and molecular medicine, 24(15), 8772-8778 (2020-07-02)
In neurodegenerative diseases such as Alzheimer's disease, Parkinson's disease, multiple sclerosis and amyotrophic lateral sclerosis, neuroinflammation can lead to blood-brain barrier (BBB) breakdown. After intravenous or intra-arterial injection into mice, endothelial progenitor cells (EPCs) home to the damaged BBB to
Reynald Thinard et al.
Pharmaceutics, 14(7) (2022-07-28)
The failures of anti-β-amyloid immunotherapies suggested that the very low fraction of injected antibodies reaching the brain parenchyma due to the filtering effect of the BBB may be a reason for the lack of therapeutic effect. However, there is no
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service