Skip to Content
Merck
All Photos(1)

Key Documents

SAB56000330

Sigma-Aldrich

Anti-SARS-CoV-2 (2019-nCov) Nucleocapsid Rabbit Monoclonal Antibody

clone RM420, affinity purified immunoglobulin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$526.00

$526.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
100 μG
$526.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$526.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

antibody form

affinity purified immunoglobulin

Quality Level

antibody product type

primary antibodies

clone

RM420, monoclonal

concentration

1 mg/mL

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Specificity

This antibody reacts to 2019-nCov Nucleocapsid Protein.

Immunogen

A recombinant 2019-nCov Nucleocapsid Protein expressed in E.coli.

Physical form

Solution in PBS

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service