Skip to Content
Merck
All Photos(1)

Key Documents

SAB5300393

Sigma-Aldrich

Monoclonal Anti-RAF1 antibody produced in mouse

clone 4G4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

CRAF, NS5, Raf-1, c-Raf

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4G4, monoclonal

form

buffered aqueous solution

mol wt

73 kDa

species reactivity

human

technique(s)

direct ELISA: 1:10,000
flow cytometry: 1:200-1:400
immunohistochemistry: 1:200-1:1,000
western blot: 1:500-1:2,000

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... RAF1(5894)

Immunogen

Purified recombinant fragment of human RAF1 expressed in E.coli.
Mouse monoclonal antibody raised against RAF1

Physical form

Purified antibody in PBS with 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiu Zhang et al.
Journal of pharmacological sciences, 126(1), 66-76 (2014-09-05)
P-glycoprotein (P-gp)-induced drug resistance is a major road block for successful cancer chemotherapy. Through phenotypic screening, the compound 2-(2-chlorophenylimino)-5-(4-dimethylaminobenzylidene) thiazolidin-4-one (CDBT) was discovered to have potent anti-tumor activity in P-gp over-expressing drug-resistant non-small-cell lung cancer (NSCLC) H460TaxR cells. Here, we
Jimin Yuan et al.
Oncogene (2018-06-23)
Although extensively studied for three decades, the molecular mechanisms that regulate the RAF/MEK/ERK kinase cascade remain ambiguous. Recent studies identified the dimerization of RAF as a key event in the activation of this cascade. Here, we show that in-frame deletions
Sudipta Maitra et al.
Molecular and cellular endocrinology, 393(1-2), 109-119 (2014-06-24)
High intra-cellular cyclic nucleotide (cAMP) ensures prophase-I arrest and prevent steroid-induced meiotic G2-M1 transition in full-grown oocytes; however, relatively less information is available for cAMP regulation of growth factor-stimulated signalling events in the oocyte model. Here using zebrafish oocytes, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service