Skip to Content
Merck
All Photos(1)

Key Documents

SAB5300321

Sigma-Aldrich

Monoclonal Anti-COX4I1 antibody produced in mouse

clone 6B3, ascites fluid

Synonym(s):

COX4, COX4-1, COX4I1, COXIV, MGC72016

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

6B3, monoclonal

mol wt

19 kDa

species reactivity

human, mouse, monkey, rat

technique(s)

direct ELISA: 1:10,000
flow cytometry: 1:200-1:400
indirect immunofluorescence: 1:200-1:1,000
western blot: 1:500-1:2,000

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COX4I1(1327)

Immunogen

Purified recombinant fragment of human COX4I1 expressed in E.coli.
Mouse monoclonal antibody raised against COX4I1

Physical form

Ascitic fluid containing 0.03% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service