Skip to Content
Merck
All Photos(1)

Key Documents

SAB5300089

Sigma-Aldrich

Monoclonal Anti-HAS1 antibody produced in mouse

clone 3E10, ascites fluid

Synonym(s):

HAS, HAS1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

3E10, monoclonal

mol wt

65 kDa

species reactivity

human

technique(s)

direct ELISA: 1:10,000
indirect immunofluorescence: 1:200-1:1,000
western blot: 1:500-1:2,000

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... HAS1(3036)

Immunogen

Purified recombinant fragment of human HAS1 expressed in E.coli.
Mouse monoclonal antibody raised against HAS1

Physical form

Ascitic fluid containing 0.03% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hanne Norsgaard et al.
Archives of dermatological research, 306(8), 719-729 (2014-07-17)
The calcipotriol/betamethasone dipropionate fixed-combination gel is widely used for topical treatment of psoriasis vulgaris. It has been hypothesized that calcipotriol counteracts glucocorticoid-induced skin atrophy which is associated with changes in the extracellular matrix (ECM). To elucidate the combined effects of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service