Skip to Content
Merck
All Photos(4)

Documents

SAB5200027

Sigma-Aldrich

Monoclonal Anti-KCNJ2 antibody produced in mouse

clone S112B-14, 1 mg/mL, purified immunoglobulin

Synonym(s):

Anti-HHBIRK1, Anti-HHIRK1, Anti-HIRK 1, Anti-IRK1, Anti-KCNJ2, Anti-Kir2.1, S112B-14

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

S112B-14, monoclonal

form

buffered aqueous glycerol solution

mol wt

antigen predicted mol wt 45 kDa

species reactivity

mouse, human, rat

concentration

1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Kcnj2(16518)

Related Categories

Specificity

Detects ~45 kDa. No cross-reactivity against Kir2.2 or Kir2.3

Immunogen

Fusion protein amino acids 41-64 and 189-428 of mouse Kir2.1, Accession Number NP_032451.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in PBS, pH 7.4, 50% glycerol, and 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polarized localization of phosphatidylserine in the endothelium regulates Kir2.1.
Ruddiman, et al.
JCI insight, 8 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service