Skip to Content
Merck
All Photos(2)

Key Documents

SAB5200001

Sigma-Aldrich

Monoclonal Anti-HSPA1A/HSPA8 antibody produced in mouse

clone BB70, 1 mg/mL, purified immunoglobulin

Synonym(s):

Anti-HSPA1, Anti-HSPA1B, Anti-Hsc54, Anti-Hsc71, Anti-Hsp70/Hsc70, Anti-LAP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

BB70, monoclonal

form

buffered aqueous glycerol solution

mol wt

antigen predicted mol wt 72-73 kDa

species reactivity

bovine, dog, beluga, guinea pig, sheep, human, pig, rat, fish, mouse, yeast, hamster, chicken, Xenopus, Drosophila, rabbit

concentration

1 mg/mL

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

chicken ... HSPA2(423504)

Specificity

Detects ~72 and ~73 kDa proteins corresponding to the molecular mass of inducible Hsp and Hsc on SDS PAGE immunoblots.

Immunogen

Chicken Hsp70/Hsp90 complex

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in PBS, pH 7.2, 50% glycerol, and 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanlong Kang et al.
Journal of medicinal chemistry, 57(4), 1188-1207 (2014-02-20)
Heat shock protein 70 (Hsp70) is an important emerging cancer target whose inhibition may affect multiple cancer-associated signaling pathways and, moreover, result in significant cancer cell apoptosis. Despite considerable interest from both academia and pharmaceutical companies in the discovery and
Hajime Shiba et al.
The Journal of experimental biology, 219(Pt 8), 1146-1153 (2016-03-06)
To elucidate the degradation process of the posterior silk gland during metamorphosis of the silkworm ITALIC! Bombyx mori, tissues collected on the 6th day after entering the 5th instar (V6), prior to spinning (PS), during spinning (SP) and after cocoon

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service