Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700514

Sigma-Aldrich

Monoclonal Anti-TCR beta antibody produced in mouse

clone MEM-262, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-TCR β (Vβ5.3-related)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MEM-262, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG2a

NCBI accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... TRB@(6957)

Related Categories

General description

The antibody MEM-262 recognizes β chains of the TCR expressed by HPB-ALL cell line [carrying V(beta5.3)] and a small subset of peripheral blood T cells.
TCR β/TCRB (T cell receptor β chain) is located on human chromosome 7q34. This gene has at least 65 rearrangeable Vβ gene segments (27 Vβ families), two Dβ segments (Dβ1 and Dβ2), two clusters of Jβ gene segments (Jβ1.1–Jβ 1.6 and Jβ2.1–Jβ2.7) and two Cβ gene segments.

Immunogen

Human thymoma cell line HPB-ALL

Application

The reagent is designed for Flow Cytometry analysis. Suggested working dilution for Flow Cytometry is 2-8 μg/mL of sample. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Biochem/physiol Actions

The complementary determining region 3 (CDR3) of the TCR β/TCRB (T cell receptor β) is essential for the identification of the major histocompatibility complex (MHC)−peptide complex.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TCRB gene rearrangements in childhood and adult precursor-B-ALL: frequency, applicability as MRD-PCR target, and stability between diagnosis and relapse
Van der Velden, et al.
Leukemia, 18(12), 1971-1971 (2004)
The T-cell receptor repertoire influences the tumor microenvironment and is associated with survival in aggressive B-cell lymphoma
Keane C, et al.
Clinical Cancer Research, 23(7), 1820-1828 (2017)
T-lineage acute lymphoblastic leukemia (T-ALL)
Raimondi S C
Atlas of Genetics and Cytogenetics in Oncology and Haematology (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service