SAB4600146
Monoclonal Anti-GFP IgG (H+L), CF™ 633 antibody produced in mouse
~1 mg/mL, clone 9F9.F9, purified immunoglobulin
About This Item
biological source
mouse
conjugate
CF™ 633 conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
9F9.F9, monoclonal
form
buffered aqueous solution
concentration
~1 mg/mL
technique(s)
indirect immunofluorescence: 1-10 μg/mL
isotype
IgG1κ
fluorescence
λex 630 nm; λem 650 nm
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Specificity
Immunogen
Features and Benefits
Physical form
Preparation Note
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service