SAB4600051
Monoclonal Anti-GFP IgG, CF™ 488A antibody produced in mouse
~1 mg/mL, clone 9F9.F9, purified immunoglobulin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
mouse
conjugate
CF™ 488A conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
9F9.F9, monoclonal
form
buffered aqueous solution
concentration
~1 mg/mL
technique(s)
indirect immunofluorescence: 1-10 μg/mL
isotype
IgG1κ
fluorescence
λex 490 nm; λem 515 nm
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Specificity
Binds GFP
Immunogen
recombinant GFP
Application
Monoclonal Anti-GFP IgG, CF™ 488A antibody produced in mouse has been used in whole-mount immunolocalization experiment.
Biochem/physiol Actions
GFP (green fluorescence protein) is a reporter molecule which is used for checking gene expression and protein localization. GFP emits green light when it is excited with UV/blue light. The GFP fluorescence remains stable and can be detected non-invasively in living cells.
Features and Benefits
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Physical form
Supplied in phosphate buffered saline with 0.05% sodium azide, 50% glycerol and 1 mg/mL bovine serum albumin.
Preparation Note
Protect from light.
Legal Information
This product is distributed by Sigma-Aldrich Co. under the authorization of Biotium, Inc. This product is covered by one or more US patents and corresponding patent claims outside the US patents or pending applications owned or licensed by Biotium, Inc. including without limitation: 12/334,387; 12/607,915; 12/699,778; 12/850,578; 61/454,484. In consideration of the purchase price paid by the buyer, the buyer is hereby granted a limited, non-exclusive, non-transferable license to use only the purchased amount of the product solely for the buyer′s own internal research in a manner consistent with the accompanying product literature. Except as expressly granted herein, the sale of this product does not grant to or convey upon the buyer any license, expressly, by implication or estoppel, under any patent right or other intellectual property right of Biotium, Inc. Buyer shall not resell or transfer this product to any third party, or use the product for any commercial purposes, including without limitation, any diagnostic, therapeutic or prophylactic uses. This product is for research use only. Any other uses, including diagnostic uses, require a separate license from Biotium, Inc. For information on purchasing a license to use this product for purposes other than research, contact Biotium, Inc., 3159 Corporate Place, Hayward, CA 94545, Tel: (510) 265-1027. Fax: (510) 265-1352. Email: btinfo@biotium.com.
6538 is a trademark of American Type Culture Collection
CF is a trademark of Biotium, Inc.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Green fluorescent protein as a reporter of gene expression and protein localization.
Biotechniques, 19, 650-655 (1995)
The Journal of biological chemistry, 288(3), 1871-1882 (2012-12-04)
Calcium (Ca(2+)) is a key second messenger in eukaryotes and regulates diverse cellular processes, most notably via calmodulin (CaM). In Arabidopsis thaliana, IQD1 (IQ67 domain 1) is the founding member of the IQD family of putative CaM targets. The 33
Veterinary research, 50(1), 51-51 (2019-06-27)
Mycoplasma hyopneumoniae, the agent of porcine enzootic pneumonia (EP), is able to persist in the lung tissue and evade destruction by the host for several weeks. To understand the mechanism of pathogen survival, phagocytic uptake of M. hyopneumoniae by primary
Cell stem cell, 24(3), 487-497 (2019-02-26)
Human brain organoid techniques have rapidly advanced to facilitate investigating human brain development and diseases. These efforts have largely focused on generating telencephalon due to its direct relevance in a variety of forebrain disorders. Despite its importance as a relay
Molecular cell, 79(1), 84-98 (2020-06-12)
Rett syndrome (RTT), mainly caused by mutations in methyl-CpG binding protein 2 (MeCP2), is one of the most prevalent intellectual disorders without effective therapies. Here, we used 2D and 3D human brain cultures to investigate MeCP2 function. We found that
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service