Skip to Content
Merck
All Photos(2)

Key Documents

SAB4504019

Sigma-Aldrich

Anti-phospho-ALK (pTyr1604) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ALK1, Anti-CD246, Anti-NBLST3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 176 kDa

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr1604)

Gene Information

human ... ALK(238)

General description

The anaplastic lymphoma kinase (ALK) gene codes for a transmembrane receptor tyrosine kinase (RTK) that has 1620 amino acids. It is a member of the insulin receptor superfamily and is usually expressed in nerve cells. ALK gene is mapped to human chromosome 2p23.2.

Immunogen

The antiserum was produced against synthesized peptide derived from human ALK around the phosphorylation site of Tyr1604.

Immunogen Range: 1570-1619

Biochem/physiol Actions

Anaplastic lymphoma kinase (ALK) has the ability to trigger Stat3 (signal transducer and activator of transcription 3) and rescue hematopoietic cells from cell death.Mutations in ALK-1 result in hereditary hemorrhagic telangiectasia (HHT), an autosomal dominant disorder of localized angiodysplasia.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anaplastic lymphoma kinase (ALK) activates Stat3 and protects hematopoietic cells from cell death
Zamo A, et al.
Oncogene, 21(7), 1038-1047 (2002)
The role of anaplastic lymphoma kinase in pediatric cancers
Takita J, et al.
Cancer Science, 108(10), 1913-1920 (2017)
A new fusion gene TPM3-ALK in anaplastic large cell lymphoma created by a (1;2)(q25;p23) translocation
Lamant L, et al.
Blood, 93(9), 3088-3095 (1999)
Assignment of transforming growth factor beta1 and beta3 and a third new ligand to the type I receptor ALK-1
Lux A, et al.
The Journal of Biological Chemistry, 274(15), 9984-9992 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service