The SNCA (synuclein α) gene codes for a 140-amino acid protein α-synuclein, that is mapped to human chromosome 4q21-23.
Immunogen
The antiserum was produced against synthesized peptide derived from human Synuclein around the phosphorylation site of Ser129.
Immunogen Range: 91-140
Application
Anti-phospho-Synuclein (pSer129) antibody has been used in immunohistochemistry.
Biochem/physiol Actions
Accumulation of α-synuclein in oligodendrocytes results in the loss of myelin and causes neurodegeneration, leading to multiple system atrophy. Synuclein-α is known to induce oligodendrocyte maturation. Mutations in the SNCA result in parkinson disease.
Features and Benefits
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Physical form
Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
FTY720/Fingolimod Reduces Synucleinopathy and Improves Gut Motility in A53T Mice: CONTRIBUTIONS OF PRO-BRAIN-DERIVED NEUROTROPHIC FACTOR (PRO-BDNF) AND MATURE BDNF
Vidal-Martinez G, et al.
The Journal of Biological Chemistry, 291(39), 20811-20821 (2016)
The Journal of biological chemistry, 291(39), 20811-20821 (2016-08-17)
Patients with Parkinson's disease (PD) often have aggregated α-synuclein (aSyn) in enteric nervous system (ENS) neurons, which may be associated with the development of constipation. This occurs well before the onset of classic PD motor symptoms. We previously found that
α-Synucleinopathy associated with G51D SNCA mutation: a link between Parkinson's disease and multiple system atrophy
Kiely AP, et al.
Acta Neuropathologica, 125(5), 753-769 (2013)
Genetic etiology of Parkinson disease associated with mutations in the SNCA, PARK2, PINK1, PARK7, and LRRK2 genes: a mutation update
Nuytemans K, et al.
Human Mutation, 31(7), 763-780 (2010)
?-Synuclein-induced myelination deficit defines a novel interventional target for multiple system atrophy.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.