Skip to Content
Merck
All Photos(9)

Key Documents

SAB4503944

Sigma-Aldrich

Anti-phospho-MYPT1 (pThr853) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-phospho-MYPT1, Phospho-MYPT1 Detection, Rabbit Anti-MYPT1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 115 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr853)

Gene Information

human ... PPP1R12A(4659)

Immunogen

The antiserum was produced against synthesized peptide derived from human MYPT1 around the phosphorylation site of Thr853.

Immunogen Range: 621-670

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vijay Pralhad Kale et al.
Pharmaceuticals (Basel, Switzerland), 16(8) (2023-08-26)
The Rho associated coiled-coil containing protein kinase (ROCK1 and ROCK2) and myotonic dystrophy-related Cdc-42 binding kinases (MRCKα and MRCKβ) are critical regulators of cell proliferation and cell plasticity, a process intimately involved in cancer cell migration and invasion. Previously, we
Feng Liu et al.
Kidney & blood pressure research, 43(5), 1425-1436 (2018-09-14)
Endothelial-to-mesenchymal transition (EndMT) of glomerular endothelial cells (GEnCs) can induce albuminuria in diabetic nephropathy. Melatonin attenuates diabetic nephropathy, but its role and mechanism in EndMT of GEnCs in diabetic nephropathy remain unknown. The effect of melatonin on EndMT induced by
Guang Feng et al.
International immunopharmacology, 72, 176-185 (2019-04-16)
Eph receptor tyrosine kinases have a wide range of biological functions and have gradually been recognized increasingly as key regulators of inflammation and injury diseases. Although previous studies suggested that EphA2 receptor may be involved in the regulation of inflammation
Juhyun Park et al.
The world journal of men's health, 37(2), 210-218 (2018-12-28)
To compare the improving effects of diabetic erectile dysfunction with two anti-glycemic agents; phlorizin and insulin. Sixty Sprague-Dawley rats were divided into four groups (n=15 in each group): normal control (C), untreated diabetic rats (D), and diabetic rats treated by
Yonglin Yu et al.
Colloids and surfaces. B, Biointerfaces, 170, 1-10 (2018-06-03)
Titanium substrates with micro/nano hierarchical features could positively mediate the osteogenesis of a titanium implant; nevertheless, the underlying molecular mechanism needs to be further revealed. In this work, we fabricated a micro/nano hierarchically structured Ti (MNT) sample and attempted to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service