Skip to Content
Merck
All Photos(1)

Key Documents

SAB4503405

Sigma-Aldrich

Anti-HAT antibody produced in rabbit

affinity isolated antibody

Synonym(s):

KAT1, histone acetyltransferase 1, histone acetyltransferase type B catalytic

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 49 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HAT1(8520)

General description

Histone acetyltransferase 1 (HAT1) is a type B histone acetyltransferase localized in the cytosol and the nucleus. The HAT1 gene is mapped on the human chromosome at 2q31.1.

Immunogen

The antiserum was produced against synthesized peptide derived from human HAT.

Immunogen Range: 331-380

Application

Anti-HAT antibody produced in rabbit has been used in western blotting (1:500) and immunofluorescence (1:250)[1].

Biochem/physiol Actions

Histone acetyltransferase 1 (HAT1) is involved in the acetylation of lysine 5- and 12- residues at the N-terminal of the newly synthesized histone H4. This enzyme is also involved in the regulation of DNA repair in the nucleus.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaohan Yang et al.
The Journal of biological chemistry, 288(25), 18271-18282 (2013-05-09)
Faithful repair of DNA double-strand breaks is vital to the maintenance of genome integrity and proper cell functions. Histone modifications, such as reversible acetylation, phosphorylation, methylation, and ubiquitination, which collectively contribute to the establishment of distinct chromatin states, play important
Hirak Kumar Barman et al.
Biochemical and biophysical research communications, 373(4), 624-630 (2008-07-08)
Amounts of soluble histones in cells are tightly regulated to ensure supplying them for the newly synthesized DNA and preventing the toxic effect of excess histones. Prior to incorporation into chromatin, newly synthesized histones H3 and H4 are highly acetylated
Josef Davidsson et al.
Epilepsy research, 81(1), 69-79 (2008-06-10)
To characterize a deletion of chromosome 2q at the molecular level in a patient suffering from severe epilepsy resembling severe myoclonic epilepsy of infancy/Dravet's syndrome (SMEI/DS) and to correlate other cases harboring deletions in the same region to morphological and
Yueqin Wang et al.
Chemosphere, 241, 125073-125073 (2019-11-07)
Microcystin-leucine arginine (MC-LR) is a variant of microcystins (MCs), which poses a serious threat to the reproductive system. Histone acetylation modification can regulate the expressions of apoptosis-related genes. However the mechanisms of histone acetylation involving MC-LR-induced apoptosis were not understood.
S V Demyanenko et al.
Molecular neurobiology, 57(7), 3219-3227 (2020-06-09)
Stroke is one of the leading reasons of human death. Ischemic penumbra that surrounds the stroke-induced infarction core is potentially salvageable, but molecular mechanisms of its formation are poorly known. Histone acetylation induces chromatin decondensation and stimulates gene expression. We

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service