Skip to Content
Merck
All Photos(3)

Documents

SAB4502827

Sigma-Aldrich

Anti-Synuclein β antibody produced in rabbit

affinity isolated antibody

Synonym(s):

β-synuclein, SYUB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 14 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SNCB(6620)

General description

Anti-Synuclein β antibody detects endogenous levels of total Synuclein β protein.
The SNCB (synuclein β) gene is mapped to the chromosome 5q35.2. Synuclein β is known to localize along with synuclein α in the presynaptic region of the brain. It contains KTKXGV repeats at its N-terminal, highly acidic and solubilizing region at its C-terminal and a non-amyloid-β component.

Immunogen

The antiserum was produced against synthesized peptide derived from human Synuclein beta.

Immunogen Range: 85-134

Biochem/physiol Actions

Synuclein β reduces the effects of synuclein α toxicity in a dose dependent manner by preventing its accumulation. Synuclein β normally, does not form fibrils.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The loss of inhibitory C-terminal conformations in disease associated P123H ?-synuclein.
Janowska MK and Baum J
Protein Science, 25(1), 286-294 (2016)
Prenatal diagnosis and molecular cytogenetic characterization of a 1.07-Mb microdeletion at 5q35.2-q35.3 associated with NSD1 haploinsufficiency and Sotos syndrome.
Chen CP
Taiwanese Journal of Obstetrics & Gynecology, 53(4), 583-587 (2014)
Lucio M Schiapparelli et al.
Cell reports, 38(4), 110287-110287 (2022-01-27)
Intercellular transfer of toxic proteins between neurons is thought to contribute to neurodegenerative disease, but whether direct interneuronal protein transfer occurs in the healthy brain is not clear. To assess the prevalence and identity of transferred proteins and the cellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service