Skip to Content
Merck
All Photos(2)

Key Documents

SAB4502549

Sigma-Aldrich

Anti-BAX antibody produced in rabbit

affinity isolated antibody

Synonym(s):

BAX, BAX protein, cytoplasmic isoform δ

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 15 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BAX(581)

General description

Anti-BAX Antibody detects endogenous levels of total BAX protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human BAX.

Immunogen Range: 80-129

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sakhila K Banu et al.
Reproductive toxicology (Elmsford, N.Y.), 68, 171-190 (2016-07-23)
Environmental contamination with hexavalent chromium (CrVI) is a growing problem both in the U.S and developing countries. CrVI is a heavy-metal endocrine disruptor; women working in Cr industries exhibit an increased incidence of premature abortion and infertility. The current study
Kirthiram K Sivakumar et al.
Reproductive toxicology (Elmsford, N.Y.), 109, 121-134 (2022-03-22)
Environmental contamination with hexavalent chromium, Cr(VI), has been increasing in the United States as well as in developing countries. Exposure to Cr(VI) predisposes the human population to various diseases, including cancer, infertility, and developmental problems in children. Previous findings from
Quanlai Zhao et al.
Molecular medicine reports, 15(4), 2280-2288 (2017-03-06)
The aim of the present study was to investigate the role of myristicin (Myr; 1‑allyl‑5‑methoxy‑3,4‑methylenedioxybenzene), an active aromatic compound isolated from nutmeg, carrot, basil, cinnamon and parsley, in hypoxia‑induced apoptosis in rat dorsal root ganglion (DRG) neurons. It was observed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service