Skip to Content
Merck
All Photos(2)

Key Documents

SAB4502244

Sigma-Aldrich

Anti-CYTL1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

C4orf4, Cytokine-like protein 1, Protein C17

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 15 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CYTL1(54360)

General description

Anti-CYTL1 antibody detects endogenous levels of total CYTL1 protein.
The CYTL1 (cytokine like 1) gene is mapped to human chromosome 4p16.2. The gene was first discovered in CD34(+) cells of the bone marrow and cord blood. The encoded protein is predominantly expressed in chondrocytes.

Immunogen

The antiserum was produced against synthesized peptide derived from human CYTL1.

Immunogen Range: 61-110

Biochem/physiol Actions

In vitro studies reveal that Cytl1 (cytokine like 1) acts as an autocrine or paracrine regulatory factor in the process of cartilage formation. CYTL1, being homologous with IL-2 (interleukin-2), is considered as a potential cytokine that is involved in regulating immune responses. CYTL1 is known to be associated with the pathogenesis of rheumatoid arthritis and osteoarthritis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tanja Vukićević et al.
Journal of the American Society of Nephrology : JASN, 30(5), 795-810 (2019-04-17)
Arginine-vasopressin (AVP) binding to vasopressin V2 receptors promotes redistribution of the water channel aquaporin-2 (AQP2) from intracellular vesicles into the plasma membrane of renal collecting duct principal cells. This pathway fine-tunes renal water reabsorption and urinary concentration, and its perturbation
Identification of CCR2-binding features in Cytl1 by a CCL2-like chemokine model.
Tomczak A
Proteins: Structure, Function, and Genetics, 79(4), 1277-1292 (2011)
Cytokine-like 1 Chemoattracts Monocytes/Macrophages via CCR2.
Wang X
Journal of Immunology, 196(10), 4090-4099 (2016)
Xiaolin Wang et al.
Oncoimmunology, 8(5), e1577126-e1577126 (2019-05-10)
CYTL1 is a novel cytokine that was first identified in CD34+ hematopoietic cells. We previously prepared recombinant CYTL1 and verified that it chemoattracted human monocytes via the CCR2/ERK pathway. It has been reported that CYTL1 plays contradictory roles in neuroblastoma
Doris Schneller et al.
Circulation research, 124(2), 243-255 (2018-12-26)
Endothelial colony forming cells (ECFCs) or late blood outgrowth endothelial cells can be isolated from human cord or peripheral blood, display properties of endothelial progenitors, home into ischemic tissues and support neovascularization in ischemic disease models. To assess the functions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service