Skip to Content
Merck
All Photos(1)

Key Documents

SAB4501648

Sigma-Aldrich

Anti-C-KIT antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CD117 antigen, Proto-oncogene tyrosine-protein kinase Kit, SCFR, c-kit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 109 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KIT(3815)

General description

Anti-C-KIT Antibody detects endogenous levels of total C-KIT protein.
KIT (proto-oncogene) is a type III receptor tyrosine kinase (RTK). It codes for a tyrosine kinase receptor (CD117). It is located on human chromosome 4q12.
The KIT (proto-oncogene receptor tyrosine kinase) gene is mapped to human chromosome 4q12. It encodes for a transmembrane glycoprotein that belongs to the receptor tyrosine kinases subclass III family.

Immunogen

The antiserum was produced against synthesized peptide derived from human c-Kit.

Immunogen Range: 688-737

Biochem/physiol Actions

KIT (proto-oncogene receptor tyrosine kinase) is considered to be an important hematopoietic growth factor receptor. It is known to exhibit tyrosine kinase activity that is associated with a variety of biologic processes, such as cell proliferation, survival, apoptosis, and motility. Downregulation of this gene is observed during the disease progression of multiple myeloma. Variation in the gene contributes to at least 2% of all melanomas and is most frequently observed in acral and mucosal melanomas. Mutations in KIT is associated with gastrointestinal stromal tumors.
KIT (proto-oncogene) participates in cell signal transduction. It also participates in the progression of hematopoietic stem cells, melanocytes, germ cells, mast cells and interstitial cells of Cajal. Mutations in KIT gene results in gastrointestinal stromal tumors.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Assessment of CD56 and CD117 Expressions at the Time of the Diagnosis in Multiple Myeloma Patients
Ceran F, et al.
Turkish journal of haematology : official journal of Turkish Society of Haematology, 34(3), 226-226 (2017)
Loss of c-KIT expression in thyroid cancer cells.
Franceschi S, et al.
PLoS ONE, 12(3) (2017)
KIT Exon 11 Codons 557?558 Deletion Mutation Promotes Liver Metastasis Through the CXCL12/CXCR4 Axis in Gastrointestinal Stromal Tumors
Wang HC, et al.
Clinical Cancer Research, 22(14), 3477-3487 (2016)
Efficacy and safety of nilotinib in patients with KIT-mutated metastatic or inoperable melanoma: final results from the global, single-arm, phase II TEAM trial
Guo J, et al.
Annals of Oncology, 28(6), 1380-1387 (2017)
KIT extracellular and kinase domain mutations in gastrointestinal stromal tumors
Lux ML, et al.
The American Journal of Pathology, 156(3), 791-795 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service