Skip to Content
Merck
All Photos(2)

Key Documents

SAB4501253

Sigma-Aldrich

Anti-GPR18 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

G-protein coupled receptor 18, GPCRW, GPR18, N-arachidonyl glycine receptor, NAGly receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 38 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GPR18(2841)

General description

Anti-GPR18 antibody detects endogenous levels of total GPR18 protein. G protein-coupled receptor 18 (GPR18) is made up of 331 amino acids. GPR18 mRNA is located in murine spleen, bone marrow, thymus, lung and cerebellar tissue. This gene is located on human chromosome 13q32.

Immunogen

The antiserum was produced against synthesized peptide derived from human GPR18.

Immunogen Range: 191-240

Biochem/physiol Actions

G protein-coupled receptor 18 (GPR18) participates in cell migration and apoptosis. It modulates mitogen-activated protein kinases (MAPK) activity and immune stimulation.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junio Dort et al.
Nature communications, 12(1), 6264-6264 (2021-10-31)
Lack of dystrophin causes muscle degeneration, which is exacerbated by chronic inflammation and reduced regenerative capacity of muscle stem cells in Duchenne Muscular Dystrophy (DMD). To date, glucocorticoids remain the gold standard for the treatment of DMD. These drugs are
D McHugh
British journal of pharmacology, 167(8), 1575-1582 (2012-05-09)
A review of what is presently known about the G protein coupled receptor GPR18 in terms of its expression and distribution, pharmacology and potential implications for central nervous system and endocannabinoid system signalling. LINKED ARTICLES This article is part of
Characterization of non-olfactory GPCRs in human sperm with a focus on GPR18
Flegel C, et al.
Scientific Reports, 6, 32255-32255 (2016)
Ceren Emre et al.
Communications biology, 5(1), 245-245 (2022-03-23)
Sustained microglial activation and increased pro-inflammatory signalling cause chronic inflammation and neuronal damage in Alzheimer's disease (AD). Resolution of inflammation follows neutralization of pathogens and is a response to limit damage and promote healing, mediated by pro-resolving lipid mediators (LMs).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service