Skip to Content
Merck
All Photos(2)

Documents

SAB4501100

Sigma-Aldrich

Anti-KANK2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Ankyrin repeat domain-containing protein 25, Matrix-remodeling-associated protein 3, SRC-1-interacting protein, SRC1-interacting protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 91 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KANK2(25959)

General description

Anti-KANK2 Antibody detects endogenous levels of total KANK2 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human KANK2.

Immunogen Range: 351-400

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qing Pei et al.
Biomolecules, 12(3) (2022-03-26)
Acute lung injury is the most common type of organ damage with high incidence and mortality in sepsis, which is a poorly understood syndrome of disordered inflammation. The aims of this study are to explore whether heat shock protein 70

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service