Skip to Content
Merck
All Photos(2)

Documents

SAB4500839

Sigma-Aldrich

Anti-FAP-1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

PTP-BAS, PTP1E, PTPL1, PTPN13, α subunit, DPPIV, FAPA, SEPRASE, fibroblast activation protein

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 87 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FAP(2191)

General description

Fibroblast activation protein 1 (FAP-1) is a membrane-associated protein tyrosine phosphatase. It is made up of an ezrin-like cytoskeleton binding domain, six PSD95/Dlg/Z-1 homology (PDZ) domains, and a leucine zipper motif. The FAP-1 gene is located on the human chromosome at 4q21.3. Anti-FAP-1 antibody detects endogenous levels of total FAP-1 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human FAP-1.

Immunogen Range: 331-380

Application

Anti-FAP-1 antibody produced in rabbit has been used in western blotting (1: 500) and immunohistochemistry (10 μg/ml).

Biochem/physiol Actions

Fibroblast activation protein 1 (FAP-1) is involved in tumorigenesis, apoptosis, cancer progression, and insulin signaling. It acts as a prognostic marker in high-grade serous ovarian cancer and breast cancer. Overexpression of the FAP-1 gene is associated with several human cancers such as hepatocellular, pancreatic, ovarian, hematological, and colon cancer. Mutations in the FAP-1 gene lead to liver and lung cancer.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoming Bai et al.
Cell communication and signaling : CCS, 21(1), 242-242 (2023-09-19)
Cancer-associated fibroblasts (CAFs) are critically involved in tumor progression by maintaining extracellular mesenchyma (ECM) production and improving tumor development. Cyclooxygenase-2 (COX-2) has been proved to promote ECM formation and tumor progression. However, the mechanisms of COX-2 mediated CAFs activation have
Vladimir N Ivanov et al.
Molecular and cellular biology, 23(10), 3623-3635 (2003-05-02)
As revealed by intracellular pools of nonactive Fas (Apo-1), export of Fas to the cell surface is often impaired in human tumors, thereby inactivating Fas ligand-mediated apoptosis. Here, we demonstrate that association with Fas-associated phosphatase 1 (FAP-1) attenuates Fas export
Yuting Jiang et al.
Environmental pollution (Barking, Essex : 1987), 266(Pt 1), 115089-115089 (2020-07-07)
Fluoride has been considered as a risk factor of cardiovascular disease due to its endothelial toxicology. However, the mechanism underlying the endothelial toxicity of fluoride has not been clearly illustrated. MiR-200c-3p was strongly linked with endothelial function and its level
J Inazawa et al.
Genomics, 31(2), 240-242 (1996-01-15)
PTPN13 is a protein tyrosine phosphatase that associates with the C-terminal negative regulatory domain in the Fas (APO-1/CD95) receptor. The PTPN13 protein contains six GLGF repeats that have been found in the rat postsynaptic density protein (PSD-95) and the Drosophila
Mohamed Hamyeh et al.
Theranostics, 10(3), 1016-1032 (2020-01-16)
Clinical data suggest that the protein tyrosine phosphatase PTPN13 exerts an anti-oncogenic effect. Its exact role in tumorigenesis remains, however, unclear due to its negative impact on FAS receptor-induced apoptosis. Methods: We crossed transgenic mice deleted for PTPN13 phosphatase activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service