Skip to Content
Merck
All Photos(2)

Key Documents

SAB4500796

Sigma-Aldrich

Anti-AKT antibody produced in rabbit

affinity isolated antibody

Synonym(s):

AKT, AKT1 kinase, C-AKT, PKB, kinase Akt1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 55 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AKT1(207)

General description

Anti-AKT Antibody detects endogenous levels of total AKT protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human Akt.

Immunogen Range: 416-465

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bingdong Jiang et al.
Oncology letters, 21(1), 14-14 (2020-11-27)
Interleukins (ILs) are involved in the occurrence and development of numerous types of cancer, and serve a critical role in the development of effective cancer therapeutics. The aim of the present study was to investigate the effect of IL-27 on
Lili Liu et al.
Bioscience reports, 40(5) (2020-05-19)
The present study aims to reveal the molecular mechanism of peroxisome proliferator-activated receptor γ (PPARγ) on sepsis-induced acute lung injury (ALI). To do that, the rat injury model was established using cecal ligation and perforation (CLP) method, followed by different
Zhixia Fan et al.
International journal of clinical and experimental pathology, 13(4), 799-809 (2020-05-02)
Psoriasis is an immune-mediated skin disease with abnormal T cells. Regulatory T cells (Treg) are a kind of cell group with immunosuppressive effects. This study aimed to explore the role of Treg cells in the pathogenesis of psoriasis and its
Zhi Teng et al.
Oncology reports, 43(2), 503-515 (2020-01-03)
Acute lymphoblastic leukemia (ALL) is a malignant hematological disease. Tanshinone IIA (Tan IIA) has antitumor activity in vitro and in vivo. The aim of the present study was to investigate the effects of Tan IIA in combination with imatinib (IM) on the proliferation, apoptosis, migration

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service