Skip to Content
Merck
All Photos(1)

Documents

SAB4500415

Sigma-Aldrich

Anti-ADORA1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

ADORA1, Adenosine receptor A1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 36 kDa

species reactivity

rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ADORA1(134)

General description

Anti-ADORA1 Antibody detects endogenous levels of total ADORA1 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human ADORA1.

Immunogen Range: 277-326

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shelly C Y Lin et al.
Frontiers in cellular neuroscience, 13, 207-207 (2019-06-04)
Purinergic signaling regulates important physiological processes and the homeostatic response to stress in the cochlea via extracellular nucleosides (adenosine) and nucleotides (ATP, UTP). Using a previously established organotypic culture model, the current study investigated the effect of purinergic P1 (adenosine)
Wenjuan Wan et al.
Molecular vision, 23, 1006-1014 (2018-02-02)
To evaluate the effects of 7-methylxanthine (7-MX) on the growth of human retinal pigment epithelium (RPE) cells and to observe the changes in the expression of adenosine receptors (ADORs) in RPE cells upon 7-MX treatment. Human RPE cells (monolayer at
Siqi Hong et al.
Cellular and molecular neurobiology (2018-05-18)
It is of great importance to explore the development of epileptogenesis, and the adenosine and adenosine kinase (ADK) system seems to play a key role in this process. The aim of this study is to explore the dynamic changes of
Alberto L Horenstein et al.
Frontiers in pharmacology, 9, 196-196 (2018-05-08)
Cytokine-induced killer (CIK) cells, a heterogeneous T cell population obtained by in vitro differentiation of peripheral blood mononuclear cells (PBMC), represent a promising immunological approach in cancer. Numerous studies have explored the role of CD38, CD39, CD203a/PC-1, and CD73 in
Victor Babich et al.
Journal of cellular physiology, 234(8), 13917-13930 (2019-01-12)
A high renal oxygen (O2 ) need is primarily associated with the renal tubular O2 consumption (VO2 ) necessary for a high rate of sodium (Na+ ) transport. Limited O2 availability leads to increased levels of adenosine, which regulates the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service