Skip to Content
Merck
All Photos(1)

Key Documents

SAB4500156

Sigma-Aldrich

Anti-PLK2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

PLK-2, Polo-like kinase 1, Serine/threonine-protein kinase SNK, Serum-inducible kinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 78 kDa

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLK2(10769)

General description

Anti-PLK2 Antibody detects endogenous levels of total PLK2 protein.
Polo-like kinases 2 (PLK2) is a member of the serine-threonine kinases family. The PLK2 gene is mapped to human chromosome 5q11.2. PLK2 comprises a C-terminal polo-box domain (PBD) devoid of catalytic activity and an N-terminal kinase domain.

Immunogen

The antiserum was produced against synthesized peptide derived from human PLK2.

Immunogen Range: 291-340

Application

Anti-PLK2 antibody produced in rabbit has been used in western blotting(1:1000) (1:5000)[1]

Biochem/physiol Actions

Polo-like kinase 2 (PLK2) participates in regulating centriole duplication, cell death, and cell cycle. It also mediates the spine-associated RapGAP (SPAR) degradation via the ubiquitin-proteasome pathway, thereby regulating synaptic plasticity. Mutations in the PLK2 gene are observed in colorectal cancers. The PLK2 gene knockdown leads to defects in centriole duplication.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhiqiang Gao et al.
Biochemistry and cell biology = Biochimie et biologie cellulaire, 99(4), 403-413 (2020-12-03)
Neuronal injury induced by cerebral ischemia poses a serious health risk globally, and there is no effective clinical therapy. This study was performed to investigate the role of transcription factor AP-2 alpha (TFAP2A) in cerebral ischemia, and the underlying mechanisms
Jaerak Chang et al.
The EMBO journal, 29(14), 2395-2406 (2010-06-10)
Control of centrosome duplication is tightly linked with the progression of the cell cycle. Recent studies suggest that the fundamental process of centriole duplication is evolutionally conserved. Here, we identified centrosomal P4.1-associated protein (CPAP), a human homologue of SAS-4, as
Association of TERT promoter mutations with telomerase expression in melanoma.
Seungjae Lee et al.
Pigment cell & melanoma research, 29(3), 391-393 (2016-03-02)
Ju Hee Kim et al.
Proteins, 83(7), 1201-1208 (2015-04-08)
Polo-like kinases (Plks) are the key regulators of cell cycle progression, the members of which share a kinase domain and a polo-box domain (PBD) that serves as a protein-binding module. While Plk1 is a promising target for antitumor therapy, Plk2
Frameshift mutation and loss of expression of PLK2, a serine/threonine kinase-encoding gene, in colorectal cancers.
Ju Hwa Lee et al.
Pathology, research and practice, 213(8), 1019-1020 (2017-07-13)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service