Skip to Content
Merck
All Photos(1)

Documents

SAB4301725

Sigma-Aldrich

Anti-PKD2L1

affinity isolated antibody

Synonym(s):

PCL, PKD2L, PKDL, TRPP3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20-1:100

accession no.

NP_057196

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PKD2L1(9033)

General description

PKD2L1 (polycystin 2 like 1, transient receptor potential cation channel) is also known as polycystin-L/ transient receptor potential polycystin-3 (TRPP3). It is expressed in brain, kidney, heart, skeletal muscle, spleen, testis, retina and in the bipolar neurons in the tongue taste buds. TRPP3 is present on the surface membrane and/or endoplasmic reticulum membrane of cells. It belongs to the PKD2 gene family. PKD2L1 is mapped to human chromosome 10q25.

Immunogen

Synthetic peptide of human PKD2L1

Biochem/physiol Actions

PKD2L1 (polycystin 2 like 1, transient receptor potential cation channel) participates in hedgehog signaling, intestinal development and sour tasting. It controls ciliary Ca2+ concentration. Mutations in PKD2 results in autosomal dominant kidney disease (ADPKD).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in pH7.4 PBS, 0.05% NaN3, 40% Glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification ofPKD2L, a HumanPKD2-Related Gene: Tissue-Specific Expression and Mapping to Chromosome 10q25.
Wu G, et al.
Genomics, 54(3), 564-568 (1998)
Regulation of TRPP3 channel function by N-terminal domain palmitoylation and phosphorylation.
Zheng W, et al.
The Journal of Biological Chemistry, 291(49), 25678-25691 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service