Skip to Content
Merck
All Photos(1)

Documents

SAB4301310

Sigma-Aldrich

Anti-phospho-PEK/PERK (pThr981) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

E2AK3, EIF2AK3, PEK, PERK

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

125 kDa

species reactivity

human

concentration

1.0 mg/mL

technique(s)

immunohistochemistry: 1:50-1:100

isotype

IgG

accession no.

NP_004827.4.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr981)

Gene Information

human ... EIF2AK3(9451)

General description

Protein kinase R (PKR)-like endoplasmic reticulum kinase (PERK)/EIF2AK3 (eukaryotic translation initiation factor 2 α kinase 3) is a type I transmembrane protein, located on ER (endoplasmic reticulum). It is a member of the eIF2α kinase subfamily. This gene is located on human chromosome 2p. EIF2AK3 has 16 exons and is made of cytoplasmic and kinase domains.

Specificity

The antibody detects endogenous levels of PEK/PERK only when phosphorylated at threonine 981.

Immunogen

Peptide sequence around phosphorylation site of threonine 981(A-R-H(p)-T-G) derived from Human PEK/PERK.

Biochem/physiol Actions

Protein kinase R (PKR)-like endoplasmic reticulum kinase (PERK) helps to detect the protein-folding imbalance created by endoplasmic reticulum (ER) stress. EIF2AK3 works as an endoplasmic reticulum (ER) stress sensor to modulate global protein synthesis. Mutation in EIF2AK3 gene results in Wolcott-rallison syndrome.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monogenic diabetes in children and young adults: Challenges for researcher, clinician and patient.
Slingerland AS.
Reviews in Endocrine & Metabolic Disorders, 7(3), 171-185 (2006)
The ER stress sensor PERK luminal domain functions as a molecular chaperone to interact with misfolded proteins.
Wang P, et al.
Acta crystallographica. Section D, Structural biology, 72(12), 1290-1297 (2016)
A functional haplotype in EIF2AK3, an ER stress sensor, is associated with lower bone mineral density.
Liu J, et al.
Journal of Bone and Mineral Research, 27(2), 331-341 (2012)
Protein kinase R-like ER kinase and its role in endoplasmic reticulum stress-decided cell fate.
Liu Z, et al.
Cell Death & Disease, 6(7), e1822-e1822 (2015)
Wolcott-Rallison Syndrome with Novel EIF2AK3 Gene Mutation.
Gurbuz F, et al.
Journal of Clinical Research in Pediatric Endocrinology, 8(4), 496-496 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service